2-Bromo-5-fluoronicotinic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC400184-250MG
APOSPC400184-250MGEA
220
EUR
APOSPC400184-250MG
2-Bromo-5-fluoronicotinic acid
2-Bromo-5-fluoronicotinic acid
Formel:
C₆H₃BrFNO₂ Molecular Weight: 219,996 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11870208 CAS-Nummer: 38186-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...