2-Bromo-5-fluoropyridin-3-ol 95%

Lieferant: Apollo Scientific
Achtung

PC400188-1G PC400188-250MG
APOSPC400188-1GEA 0 EUR
APOSPC400188-1G APOSPC400188-250MG
2-Bromo-5-fluoropyridin-3-ol 95%
2-Bromo-5-fluoropyridin-3-ol
Formel: C₅H₃BrFNO
Lagertemperatur: Raumtemperatur
MDL: MFCD11519042
CAS-Nummer: 1093758-87-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR