2-(Difluoromethyl)-5-nitropyridin-4-ol

Lieferant: Apollo Scientific
Achtung

PC400192-1G PC400192-250MG PC400192-5G
APOSPC400192-1GEA 0 EUR
APOSPC400192-1G APOSPC400192-250MG APOSPC400192-5G
2-(Difluoromethyl)-5-nitropyridin-4-ol
2-(Difluoromethyl)-5-nitropyridin-4-ol
Formel: C₆H₄F₂N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD25481266
CAS-Nummer: 1805314-41-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR