3-(Trifluormethoxy)benzolsulfonamid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Trifluormethoxy)benzolsulfonsäureamid

PC4004-1G PC4004-25G PC4004-5G
APOSPC4004-1GEA 29 EUR
APOSPC4004-1G APOSPC4004-25G APOSPC4004-5G
3-(Trifluormethoxy)benzolsulfonamid
3-(Trifluormethoxy)benzolsulfonamid
Formel: C₇H₆F₃NO₃S
Molecular Weight: 241.19 g/mol
Schmelzpunkt: 100…101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094233
CAS-Nummer: 503179-70-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR