Ethyl 3-{[(trifluoromethyl)sulphonyl]oxy}furo[2,3-c]pyridine-2-carboxylate

Lieferant: Apollo Scientific

Synonyme: 2-(Ethoxycarbonyl)-3-{[(trifluoromethyl)sulphonyl]oxy}furo[2,3-c]pyridine, 2-(Ethoxycarbonyl)furo[2,3-c]pyridin-3-yl trifluoromethanesulphonate

PC400501-250MG
APOSPC400501-250MGEA 466 EUR
APOSPC400501-250MG
Ethyl 3-{[(trifluoromethyl)sulphonyl]oxy}furo[2,3-c]pyridine-2-carboxylate
Ethyl 3-{[(trifluoromethyl)sulphonyl]oxy}furo[2,3-c]pyridine-2-carboxylate
Formel: C₁₁H₈F₃NO₆S
Molecular Weight: 339,245 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22690758
CAS-Nummer: 927804-63-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR