3-Amino-5-(1-fluoroethyl)-1H-pyrazole
Lieferant: Apollo Scientific
Synonyme:
5-(1-Fluoroethyl)-1H-pyrazol-3-amine
PC400502-250MG
APOSPC400502-250MGEA
742
EUR
APOSPC400502-250MG
3-Amino-5-(1-fluoroethyl)-1H-pyrazole
3-Amino-5-(1-fluoroethyl)-1H-pyrazole
Formel:
C₅H₈FN₃ Molecular Weight: 129,136 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22690772 CAS-Nummer: 1451391-06-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...