1-(Azetidin-3-yl)-4,4-difluoropiperidine

Lieferant: Apollo Scientific
Achtung

PC400503-250MG
APOSPC400503-250MGEA 844 EUR
APOSPC400503-250MG
1-(Azetidin-3-yl)-4,4-difluoropiperidine
1-(Azetidin-3-yl)-4,4-difluoropiperidine
Formel: C₈H₁₄F₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22123272
CAS-Nummer: 1257293-83-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR