4-(3-Fluoroazetidin-1-yl)piperidine dihydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluoro-1-(piperidin-4-yl)azetane dihydrochloride, 3-Fluoro-1-(piperidin-4-yl)azetidine dihydrochloride

PC400508-250MG
APOSPC400508-250MGEA 696 EUR
APOSPC400508-250MG
4-(3-Fluoroazetidin-1-yl)piperidine dihydrochloride
4-(3-Fluoroazetidin-1-yl)piperidine dihydrochloride
Formel: C₈H₁₅FN₂·2ClH
Molecular Weight: 231,138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22416295
CAS-Nummer: 1403767-21-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR