4-(5-Fluoro-2-hydrazinopyrimidin-4-yl)morpholine

Lieferant: Apollo Scientific

Synonyme: [5-Fluoro-4-(morpholin-4-yl)pyrimidin-2-yl]hydrazine, 5-Fluoro-2-hydrazino-4-(morpholin-4-yl)pyrimidine

PC400510-1G
APOSPC400510-1GEA 677 EUR
APOSPC400510-1G
4-(5-Fluoro-2-hydrazinopyrimidin-4-yl)morpholine
4-(5-Fluoro-2-hydrazinopyrimidin-4-yl)morpholine
Formel: C₈H₁₂FN₅O
Molecular Weight: 213.212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00181489
CAS-Nummer: 118121-89-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR