5-Fluoro-1H-indole-4-boronic acid, pinacol ester

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole, 2-(5-Fluoro-1H-indol-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PC400511-500MG
APOSPC400511-500MGEA 1020 EUR
APOSPC400511-500MG
5-Fluoro-1H-indole-4-boronic acid, pinacol ester
5-Fluoro-1H-indole-4-boronic acid, pinacol ester
Formel: C₁₄H₁₇BFNO₂
Molecular Weight: 261,1 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD16987816
CAS-Nummer: 1072009-08-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR