N'-(4-Fluorobenzyl)ethane-1,2-diamine, N-BOC protected

Lieferant: Apollo Scientific

Synonyme: tert-Butyl {2-[(4-fluorobenzyl)amino]ethyl}carbamate

PC400513-1G PC400513-500MG
APOSPC400513-1GEA 649 EUR
APOSPC400513-1G APOSPC400513-500MG
N'-(4-Fluorobenzyl)ethane-1,2-diamine, N-BOC protected
N'-(4-Fluorobenzyl)ethane-1,2-diamine, N-BOC protected
Formel: C₁₄H₂₁FN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22690784
CAS-Nummer: 519172-79-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR