3-Bromo-2-chloro-6-fluorobenzaldehyde

Lieferant: Apollo Scientific
Achtung

PC400515-1G PC400515-5G PC400515-25G
APOSPC400515-1GEA 36.4 EUR
APOSPC400515-1G APOSPC400515-5G APOSPC400515-25G
3-Bromo-2-chloro-6-fluorobenzaldehyde
3-Bromo-2-chloro-6-fluorobenzaldehyde
Formel: C₇H₃BrClFO
Lagertemperatur: Raumtemperatur
MDL: MFCD11110260
CAS-Nummer: 1114809-11-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR