3-(Fluoromethyl)piperidine hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

PC400517-250MG
APOSPC400517-250MGEA 0 EUR
APOSPC400517-250MG
3-(Fluoromethyl)piperidine hydrochloride 95%
3-(Fluoromethyl)piperidine hydrochloride
Formel: C₆H₁₃ClFN
Lagertemperatur: Raumtemperatur
MDL: MFCD17015069
CAS-Nummer: 1241725-60-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR