5-Amino-6-chloro-4-fluoro-1H-benzimidazole

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-4-fluoro-1H-benzimidazol-5-amine

PC400519-250MG PC400519-500MG
APOSPC400519-250MGEA 524 EUR
APOSPC400519-250MG APOSPC400519-500MG
5-Amino-6-chloro-4-fluoro-1H-benzimidazole
5-Amino-6-chloro-4-fluoro-1H-benzimidazole
Formel: C₇H₅ClFN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD22690795
CAS-Nummer: 1426290-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR