Methyl 5,5-difluoropiperidine-3-carboxylate, N-BOC protected
Lieferant: Apollo Scientific
|
Achtung
|
PC400520-1G
APOSPC400520-1GEA
0
EUR
APOSPC400520-1G
Methyl 5,5-difluoropiperidine-3-carboxylate, N-BOC protected
Methyl 5,5-difluoropiperidine-3-carboxylate, N-BOC protected
Formel:
C₁₂H₁₉F₂NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18433629 CAS-Nummer: 1255667-06-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...