4-Fluoropiperidine-4-carboxylic acid, N-BOC protected 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(tert-Butoxycarbonyl)-4-carboxy-4-fluoropiperidine, 1-Boc-4-fluoro-4-piperidinecarboxylic Acid, 1-(tert-Butoxycarbonyl)-4-fluoropiperidine-4-carboxylic acid
PC400525-1G
APOSPC400525-1GEA
165
EUR
APOSPC400525-1G
4-Fluoropiperidine-4-carboxylic acid, N-BOC protected 98%
4-Fluoropiperidine-4-carboxylic acid, N-BOC protected
Formel:
C₁₁H₁₈FNO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08062530 CAS-Nummer: 614731-04-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...