6-Fluoro-1H-indole-4-boronic acid, pinacol ester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole, 2-(6-Fluoro-1H-indol-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PC400528-250MG
APOSPC400528-250MGEA
197
EUR
APOSPC400528-250MG
6-Fluoro-1H-indole-4-boronic acid, pinacol ester
6-Fluoro-1H-indole-4-boronic acid, pinacol ester
Formel:
C₁₄H₁₇BFNO₂ Molecular Weight: 261,1 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD16987817 CAS-Nummer: 955978-85-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...