4-Fluoro-1,4'-bipiperidine hydrochloride

Lieferant: Apollo Scientific

PC400530-250MG
APOSPC400530-250MGEA 633 EUR
APOSPC400530-250MG
4-Fluoro-1,4'-bipiperidine hydrochloride
4-Fluoro-1,4'-bipiperidine hydrochloride
Formel: C₁₀H₁₉FN₂·ClH
Molecular Weight: 222.731 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23097075
CAS-Nummer: 1426290-04-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR