3-Bromo-2-chloro-4,5-difluorobenzoic acid

Lieferant: Apollo Scientific
Achtung

PC400531-500MG
APOSPC400531-500MGEA 597 EUR
APOSPC400531-500MG
3-Bromo-2-chloro-4,5-difluorobenzoic acid
3-Bromo-2-chloro-4,5-difluorobenzoic acid
Formel: C₇H₂BrClF₂O₂
Molecular Weight: 271,443 g/mol
Schmelzpunkt: 137…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD22690841
CAS-Nummer: 112062-69-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR