4-Amino-2-methyl-6-(trifluoromethyl)pyrimidine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methyl-6-(trifluoromethyl)pyrimidin-4-amine, 4-Amino-2-methyl-6-(trifluoromethyl)-1,3-diazine

PC400535-500MG
APOSPC400535-500MGEA 611 EUR
APOSPC400535-500MG
4-Amino-2-methyl-6-(trifluoromethyl)pyrimidine
4-Amino-2-methyl-6-(trifluoromethyl)pyrimidine
Formel: C₆H₆F₃N₃
Molecular Weight: 177,127 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09909768
CAS-Nummer: 54518-10-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR