(1S,2S)-2-Fluorocyclopropanecarboxylic acid 95+%

Lieferant: Apollo Scientific
Achtung

PC400537-1G PC400537-250MG
APOSPC400537-1GEA 0 EUR
APOSPC400537-1G APOSPC400537-250MG
(1S,2S)-2-Fluorocyclopropanecarboxylic acid 95+%
(1S,2S)-2-Fluorocyclopropanecarboxylic acid
Formel: C₄H₅FO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD19237459
CAS-Nummer: 127199-14-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR