(1R,2S)-2-Fluorocyclopropanecarboxylic acid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC400539-1G
PC400539-250MG
APOSPC400539-1GEA
0
EUR
APOSPC400539-1G
APOSPC400539-250MG
(1R,2S)-2-Fluorocyclopropanecarboxylic acid 97%
(1R,2S)-2-Fluorocyclopropanecarboxylic acid
Formel:
C₄H₅FO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD19237658 CAS-Nummer: 167073-08-7 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...