(1R,2S)-2-Fluorocyclopropanecarboxylic acid 97%

Lieferant: Apollo Scientific
Gefahr

PC400539-1G PC400539-250MG
APOSPC400539-1GEA 0 EUR
APOSPC400539-1G APOSPC400539-250MG
(1R,2S)-2-Fluorocyclopropanecarboxylic acid 97%
(1R,2S)-2-Fluorocyclopropanecarboxylic acid
Formel: C₄H₅FO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD19237658
CAS-Nummer: 167073-08-7
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR