Racemic cis-2-Fluorocyclopropanecarboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

PC400540-1G PC400540-5G
APOSPC400540-1GEA 0 EUR
APOSPC400540-1G APOSPC400540-5G
Racemic cis-2-Fluorocyclopropanecarboxylic acid 95%
Racemic cis-2-Fluorocyclopropanecarboxylic acid
Formel: C₄H₅FO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04972869
CAS-Nummer: 105919-34-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR