tert-Butyl 4-(4-bromo-3-(difluoromethyl)-1H-pyrazol-1-yl)piperidine-1-carboxylate

Lieferant: Apollo Scientific

PC400543-1G
APOSPC400543-1GEA 1200 EUR
APOSPC400543-1G
tert-Butyl 4-(4-bromo-3-(difluoromethyl)-1H-pyrazol-1-yl)piperidine-1-carboxylate
tert-Butyl 4-(4-bromo-3-(difluoromethyl)-1H-pyrazol-1-yl)piperidine-1-carboxylate
Formel: C₁₄H₂₀BrF₂N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD23704681
CAS-Nummer: 1449117-47-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR