4-Brom-3-(trifluormethyl)-1H-pyrazol
Lieferant: Apollo Scientific
Synonyme:
4-Brom-3-(trifluormethyl)-1H-pyrazol
PC400544-1G
APOSPC400544-1GEA
1030
EUR
APOSPC400544-1G
4-Brom-3-(trifluormethyl)-1H-pyrazol
4-Bromo-3-(trifluoromethyl)-1H-pyrazole
Formel:
C₄H₂BrF₃N₂ Molecular Weight: 214,97 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04969230 CAS-Nummer: 19968-17-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...