(4-Bromo-3-fluorophenyl)methanamine hydrochloride
Lieferant: Apollo Scientific
PC400545-1G
APOSPC400545-1GEA
68.3
EUR
APOSPC400545-1G
(4-Bromo-3-fluorophenyl)methanamine hydrochloride
(4-Bromo-3-fluorophenyl)methanamine hydrochloride
Formel:
C₇H₇BrFN·ClH Molecular Weight: 240,501 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12546113 CAS-Nummer: 1214342-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...