(3-Fluoro-4-(2-fluoropyridin-4-yl)phenyl)methanamine hydrochloride

Lieferant: Apollo Scientific

PC400546-1G
APOSPC400546-1GEA 939 EUR
APOSPC400546-1G
(3-Fluoro-4-(2-fluoropyridin-4-yl)phenyl)methanamine hydrochloride
(3-Fluoro-4-(2-fluoropyridin-4-yl)phenyl)methanamine hydrochloride
Formel: C₁₂H₁₀F₂N₂·ClH
Molecular Weight: 256,679 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23704691
CAS-Nummer: 1449117-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR