3-Fluoro-2-trifluoromethylisonicotinic acid
Lieferant: Apollo Scientific
Synonyme:
3-Fluoro-2-(trifluoromethyl)isonicotinic acid
PC400548-1G
APOSPC400548-1GEA
481
EUR
APOSPC400548-1G
3-Fluoro-2-trifluoromethylisonicotinic acid
3-Fluoro-2-trifluoromethylisonicotinic acid
Formel:
C₇H₃F₄NO₂ Molecular Weight: 209,1 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08437597 CAS-Nummer: 886510-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...