tert-Butyl (4-(difluoromethyl)pyridin-2-yl)carbamate

Lieferant: Apollo Scientific

PC400549-1G
APOSPC400549-1GEA 1430 EUR
APOSPC400549-1G
tert-Butyl (4-(difluoromethyl)pyridin-2-yl)carbamate
tert-Butyl (4-(difluoromethyl)pyridin-2-yl)carbamate
Formel: C₁₁H₁₄F₂N₂O₂
Molecular Weight: 244,238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26404019
CAS-Nummer: 1419221-63-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR