4-Chloro-6-fluoroquinazoline
Lieferant: Apollo Scientific
PC400552-1G
APOSPC400552-1GEA
59.6
EUR
APOSPC400552-1G
4-Chloro-6-fluoroquinazoline
4-Chloro-6-fluoroquinazoline
Formel:
C₈H₄ClFN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07776652 CAS-Nummer: 16499-61-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...