N-Benzyloxycarbonyl-3-fluoro-4-morpholinoaniline
Lieferant: Apollo Scientific
Synonyme:
N-benzyloxycarbonyl-3-fluoro-4-morpholinoaniline, (3-Fluoro-4-morpholinophenyl)carbamic acid Benzyl ester, Benzyl (3-Fluoro-4-morpholinophenyl)carbamate
PC400553-1G
APOSPC400553-1GEA
1110
EUR
APOSPC400553-1G
N-Benzyloxycarbonyl-3-fluoro-4-morpholinoaniline
N-Benzyloxycarbonyl-3-fluoro-4-morpholinoaniline
Formel:
C₁₈H₁₉FN₂O₃ Molecular Weight: 330,359 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06658240 CAS-Nummer: 168828-81-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...