(R)-3-Chloro-2-((4-(trifluoromethoxy)benzyl)oxy)propyl 4-methoxybenzoate

Lieferant: Apollo Scientific

PC400554-1G
APOSPC400554-1GEA 715 EUR
APOSPC400554-1G
(R)-3-Chloro-2-((4-(trifluoromethoxy)benzyl)oxy)propyl 4-methoxybenzoate
(R)-3-Chloro-2-((4-(trifluoromethoxy)benzyl)oxy)propyl 4-methoxybenzoate
Formel: C₁₉H₁₈ClF₃O₅
Molecular Weight: 418,791 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26404030
CAS-Nummer: 1253202-35-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR