(4-Chlorophenyl)(1-fluorocyclopropyl)methanone
Lieferant: Apollo Scientific
PC400555-1G
APOSPC400555-1GEA
709
EUR
APOSPC400555-1G
(4-Chlorophenyl)(1-fluorocyclopropyl)methanone
(4-Chlorophenyl)(1-fluorocyclopropyl)methanone
Formel:
C₁₀H₈ClFO Molecular Weight: 198,621 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13172916 CAS-Nummer: 103543-60-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...