Ethyl 2-chloro-2-fluorocyclopropanecarboxylate

Lieferant: Apollo Scientific

PC400557-1G
APOSPC400557-1GEA 1030 EUR
APOSPC400557-1G
Ethyl 2-chloro-2-fluorocyclopropanecarboxylate
Ethyl 2-chloro-2-fluorocyclopropanecarboxylate
Formel: C₆H₈ClFO₂
Molecular Weight: 166,578 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26404044
CAS-Nummer: 155051-93-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR