7-Methoxy-N-(3-(trifluoromethyl)phenyl)quinoline-4-carboxamide

Lieferant: Apollo Scientific

PC400558-1G
APOSPC400558-1GEA 1030 EUR
APOSPC400558-1G
7-Methoxy-N-(3-(trifluoromethyl)phenyl)quinoline-4-carboxamide
7-Methoxy-N-(3-(trifluoromethyl)phenyl)quinoline-4-carboxamide
Formel: C₁₈H₁₃F₃N₂O₂
Molecular Weight: 346,303 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26404046
CAS-Nummer: 1624262-18-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR