3-Fluoro-5-methoxypyridine-2-sulphonyl chloride

Lieferant: Apollo Scientific

PC400561-1G
APOSPC400561-1GEA 1030 EUR
APOSPC400561-1G
3-Fluoro-5-methoxypyridine-2-sulphonyl chloride
3-Fluoro-5-methoxypyridine-2-sulphonyl chloride
Formel: C₆H₅ClFNO₃S
Molecular Weight: 225,625 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18412228
CAS-Nummer: 1261870-77-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR