3,5-Difluoropyridine-2-sulphonyl chloride

Lieferant: Apollo Scientific

PC400562-1G
APOSPC400562-1GEA 1030 EUR
APOSPC400562-1G
3,5-Difluoropyridine-2-sulphonyl chloride
3,5-Difluoropyridine-2-sulphonyl chloride
Formel: C₅H₂ClF₂NO₂S
Molecular Weight: 213,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18411876
CAS-Nummer: 1261587-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR