(2-Bromo-1-fluoroethyl)benzene
Lieferant: Apollo Scientific
PC400563-1G
APOSPC400563-1GEA
873
EUR
APOSPC400563-1G
(2-Bromo-1-fluoroethyl)benzene
(2-Bromo-1-fluoroethyl)benzene
Formel:
C₈H₈BrF Lagertemperatur: Raumtemperatur |
MDL:
MFCD13689011 CAS-Nummer: 1786-36-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...