α-Fluorostyrene

Lieferant: Apollo Scientific
Achtung

Synonyme: (1-Fluoroethenyl)benzene, (1-Fluorovinyl)benzene

PC400564-1G
APOSPC400564-1GEA 1030 EUR
APOSPC400564-1G
α-Fluorostyrene
α-Fluorostyrene
Formel: C₈H₇F
Molecular Weight: 122,14 g/mol
Siedepunkt: 45…46 °C (14 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD27501035
CAS-Nummer: 696-31-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR