Methyl 3-fluoro-5-iodo-4-methylbenzoate

Lieferant: Apollo Scientific

PC400565-1G
APOSPC400565-1GEA 452 EUR
APOSPC400565-1G
Methyl 3-fluoro-5-iodo-4-methylbenzoate
Methyl 3-fluoro-5-iodo-4-methylbenzoate
Formel: C₉H₈FIO₂
Molecular Weight: 294,061 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24107118
CAS-Nummer: 861905-21-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR