(2,2-Dichloro-1-fluorocyclopropyl)benzene

Lieferant: Apollo Scientific

PC400566-1G
APOSPC400566-1GEA 1350 EUR
APOSPC400566-1G
(2,2-Dichloro-1-fluorocyclopropyl)benzene
(2,2-Dichloro-1-fluorocyclopropyl)benzene
Formel: C₉H₇Cl₂F
Lagertemperatur: Raumtemperatur
MDL: MFCD27501036
CAS-Nummer: 502763-69-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR