6-Bromo-2-fluoro-3-methoxypyridine
Lieferant: Apollo Scientific
|
Achtung
|
PC400567-1G
APOSPC400567-1GEA
287
EUR
APOSPC400567-1G
6-Bromo-2-fluoro-3-methoxypyridine
6-Bromo-2-fluoro-3-methoxypyridine
Formel:
C₆H₅BrFNO Schmelzpunkt: 58…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04112568 CAS-Nummer: 850142-73-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...