7,7-Difluorotetrahydropyrrolo[1,2-a]pyrazine-1,3(2H,4H)-dione
Lieferant: Apollo Scientific
PC400568-1G
APOSPC400568-1GEA
1030
EUR
APOSPC400568-1G
7,7-Difluorotetrahydropyrrolo[1,2-a]pyrazine-1,3(2H,4H)-dione
7,7-Difluorotetrahydropyrrolo[1,2-a]pyrazine-1,3(2H,4H)-dione
Formel:
C₇H₈F₂N₂O₂ Molecular Weight: 190,147 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27501044 CAS-Nummer: 1624260-16-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...