1-[(1,1-Dimethylethyl)dimethylsilyl]-5-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-Indole
Lieferant: Apollo Scientific
PC400569-1G
APOSPC400569-1GEA
1270
EUR
APOSPC400569-1G
1-[(1,1-Dimethylethyl)dimethylsilyl]-5-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-Indole
1-[(1,1-Dimethylethyl)dimethylsilyl]-5-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-Indole
Formel:
C₂₀H₃₁BFNO₂Si Molecular Weight: 375,361 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22690780 CAS-Nummer: 1148004-02-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...