2-(Trifluoromethyl)pyrimidin-4-amine 95+%

Lieferant: Apollo Scientific

PC400571-250MG
APOSPC400571-250MGEA 128 EUR
APOSPC400571-250MG
2-(Trifluoromethyl)pyrimidin-4-amine 95+%
2-(Trifluoromethyl)pyrimidin-4-amine
Formel: C₅H₄F₃N₃
Molecular Weight: 163,101 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08436606
CAS-Nummer: 672-42-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR