4-Amino-6-methyl-2-(trifluoromethyl)pyrimidine
Lieferant: Apollo Scientific
|
Achtung
|
PC400572-1G
APOSPC400572-1GEA
937
EUR
APOSPC400572-1G
4-Amino-6-methyl-2-(trifluoromethyl)pyrimidine
4-Amino-6-methyl-2-(trifluoromethyl)pyrimidine
Formel:
C₆H₆F₃N₃ Schmelzpunkt: 173…174 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08436605 CAS-Nummer: 4571-65-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...