PA-824 98+%

Lieferant: Apollo Scientific

Synonyme: (S)-2-Nitro-6-[[4-(trifluoromethoxy)benzyl]oxy]-6,7-dihydro-5H-imidazo[2,1-b][1,3]oxazine, Pretomanid, (S)-2-Nitro-6-((4-(trifluoromethoxy)benzyl)oxy)-6,7-dihydro-5H-imidazo[2,1-b][1,3]oxazine

PC400580-100MG
APOSPC400580-100MGEA 469 EUR
APOSPC400580-100MG
PA-824 98+%
PA-824
Formel: C₁₄H₁₂F₃N₃O₅
Molecular Weight: 359,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06809939
CAS-Nummer: 187235-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR