2-Fluoro-2-phenylcyclopropanecarboxylic acid
Lieferant: Apollo Scientific
PC400596-1G
APOSPC400596-1GEA
1090
EUR
APOSPC400596-1G
2-Fluoro-2-phenylcyclopropanecarboxylic acid
2-Fluoro-2-phenylcyclopropanecarboxylic acid
Formel:
C₁₀H₉FO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09030645 CAS-Nummer: 914221-42-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...