2,2,2-Trifluoroacetic acid compound with 3-(2-(methylamino)ethyl)benzoic acid

Lieferant: Apollo Scientific

PC400598-1G
APOSPC400598-1GEA 719 EUR
APOSPC400598-1G
2,2,2-Trifluoroacetic acid compound with 3-(2-(methylamino)ethyl)benzoic acid
2,2,2-Trifluoroacetic acid compound with 3-(2-(methylamino)ethyl)benzoic acid
Formel: C₁₀H₁₃NO₂·CF₃CO₂H
Molecular Weight: 293.239 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27978439
CAS-Nummer: 1624261-41-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR